Norcampher ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,5-Methanocyclohexanon, 2-Norbornanon, 2-Oxonorbornan, Bicyclo[2.2.1]-2-heptanon
L01491.06
L01491.14
L01491.06EA
38.5
EUR
L01491.06
L01491.14
Norcampher ≥98%
Norcampher
Purity: 98+%
CAS No.: 497-38-1
Molecular Formula: C7H10O
Formula Weight: 110.16
Flash Point: 33°(91°F)
Melting Point: 92-97°
Boiling Point: 172-174°
Beilstein Reference: 1209657
UN No.: 1325
MDL No.: MFCD00074823
Formel:
C₇H₁₀O Molecular Weight: 110.16 Siedepunkt: 172…174 °C Schmelzpunkt: 92…97 °C Dichte: 1.03 Flammpunkt: 33 °C (91 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00074823 CAS-Nummer: 497-38-1 EINECS: 207-846-1 UN: 1325 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...