(2-Butyl)triphenylphosphoniumbromid 96%
Lieferant: Thermo Scientific
|
Achtung
|
L01532.06
L01532.06EA
65.7
EUR
L01532.06
(2-Butyl)triphenylphosphoniumbromid 96%
(2-Butyl)triphenylphosphoniumbromid
Purity: 96%
CAS No.: 3968-92-1
Molecular Formula: C22H24BrP
Formula Weight: 399.32
Melting Point: 227-237°
Beilstein Reference: 3580097
Formel:
C₂₂H₂₄BrP Molecular Weight: 399,32 g/mol Schmelzpunkt: 227…237 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00050236 CAS-Nummer: 3968-92-1 EINECS: 223-587-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...