2-Iodcumol 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Iodisopropylbenzol, 2-(2-Iodphenyl)propan

L01533.06
L01533.06EA 46.4 EUR
L01533.06
2-Iodcumol 95%
2-Iodcumol

Purity: 95% CAS No.: 19099-54-8 Molecular Formula: C9H11I Formula Weight: 246.09 Refractive Index: 1.5830 Odour: Aromatic Boiling Point: 110°/20mm Beilstein Reference: 2351872 MDL No.: MFCD00013704

Formel: C₉H₁₁I
Molecular Weight: 246,09 g/mol
Siedepunkt: 110 °C (20 mmHg)
Dichte: 1,54
Lagertemperatur: Raumtemperatur
MDL: MFCD00013704
CAS-Nummer: 19099-54-8
EINECS: 242-819-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR