Isoxazol ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
L01607.06
L01607.06EA
44.9
EUR
L01607.06
Isoxazol ≥99%
Isoxazole
Purity: 99%
CAS No.: 288-14-2
Molecular Formula: C3H3NO
Formula Weight: 69.06
Flash Point: 8°(46°F)
Refractive Index: 1.4270
Boiling Point: 93-95°
Beilstein Reference: 103773
UN No.: 1993
MDL No.: MFCD00003149
Formel:
C₃H₃NO Molecular Weight: 69,06 g/mol Siedepunkt: 93…95 °C Dichte: 1,078 Flammpunkt: 8 °C (46 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003149 CAS-Nummer: 288-14-2 EINECS: 206-018-7 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...