3-Methyl-1H-pyrazol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L01618.06
L01618.14
L01618.06EA
27.1
EUR
L01618.06
L01618.14
3-Methyl-1H-pyrazol ≥97%
Purity: 97%
CAS No.: 1453-58-3
Molecular Formula: C4H6N2
Formula Weight: 82.11
Flash Point: 93°(199°F)
Refractive Index: 1.4960
Boiling Point: 203-205°
Beilstein Reference: 1454
MDL No.: MFCD00005240
Formel:
C₄H₆N₂ Siedepunkt: 203…205 °C Dichte: 1,020 Flammpunkt: 93 °C (199 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005240 CAS-Nummer: 1453-58-3 EINECS: 215-925-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...