2-Chlor-4,5-xylenol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Chloro-4,5-dimethylphenol, 4-Chloro-5-hydroxy-2-xylene, 6-Chloro-3,4-xylenol

L01627.06
L01627.06EA 45.2 EUR
L01627.06
2-Chlor-4,5-xylenol ≥98%
2-Chlor-4,5-xylenol

Purity: 98% CAS No.: 1124-04-5 Molecular Formula: C8H9ClO Formula Weight: 156.61 Melting Point: 70-72° Beilstein Reference: 2042057

Formel: C₈H₉ClO
Molecular Weight: 156,61 g/mol
Schmelzpunkt: 70…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002173
CAS-Nummer: 1124-04-5
EINECS: 214-386-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR