5-Hydroxymethyluracil 98%

Lieferant: Thermo Fisher Scientific

Synonyme: 5-(Hydroxymethyl)pyrimidine-2,4(1H,3H)-dione

L01682.03 L01682.06
L01682.03EA 47.7 EUR
L01682.03 L01682.06
5-Hydroxymethyluracil 98%
5-Hydroxymethyluracil

Purity: 98% CAS No.: 4433-40-3 Molecular Formula: C5H6N2O3 Formula Weight: 142.12 Melting Point: 300° Beilstein Reference: 125482 MDL No.: MFCD00006070

Formel: C₅H₆N₂O₃
Molecular Weight: 142.114 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006070
CAS-Nummer: 4433-40-3
EINECS: 224-636-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR