Pyrrolidin-1-ylacetonitrile 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-(Pyrrolidin-1-yl)acetonitrile

L01737.06
L01737.06EA 55 EUR
L01737.06
Pyrrolidin-1-ylacetonitrile 97%
Pyrrolidin-1-ylacetonitrile

Purity: 97% CAS No.: 29134-29-0 Molecular Formula: C6H10N2 Formula Weight: 110.16 Refractive Index: 1.4563 Beilstein Reference: 106157 UN No.: UN3276 MDL No.: MFCD00014098

Formel: C₆H₁₀N₂
Molecular Weight: 110.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00014098
CAS-Nummer: 29134-29-0
EINECS: 249-463-2
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR