α-Fluorphenylessigsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Fluoro-2-phenylacetic acid, alpha-Fluorophenylacetic acid
L01745.04
L01745.MC
L01745.ME
L01745.04EA
188
EUR
L01745.04
L01745.MC
L01745.ME
α-Fluorphenylessigsäure ≥98%
α-Fluorphenylessigsäure
Purity: 98%
CAS No.: 1578-63-8
Molecular Formula: C6H5CHFCO2H
Formula Weight: 154.14
Melting Point: 78-82°
Beilstein Reference: 2087378
UN No.: 2811
MDL No.: MFCD00004221
Formel:
C₈H₇FO₂ Molecular Weight: 154.14 g/mol Schmelzpunkt: 78…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004221 CAS-Nummer: 1578-63-8 EINECS: 216-420-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...