Dicyclopropylmethanol 97%

Lieferant: Thermo Fisher Scientific

L01816.03 L01816.06
L01816.03EA 44.9 EUR
L01816.03 L01816.06
Dicyclopropylmethanol 97%
Dicyclopropylmethanol

Purity: 97% CAS No.: 14300-33-5 Molecular Formula: C7H12O Formula Weight: 112.17 Flash Point: 110°(230°F) Refractive Index: 1.4650 Boiling Point: 99-100°/12mm Beilstein Reference: 2203215 MDL No.: MFCD00019249

Formel: C₇H₁₂O
Siedepunkt: 99…100 °C (12 mmHg)
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00019249
CAS-Nummer: 14300-33-5
EINECS: 238-236-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR