9-Hydroxyfluoren ≥98%
Lieferant: Thermo Scientific
Synonyme:
9-Fluorenol
L01917.14
L01917.14EA
52.4
EUR
L01917.14
9-Hydroxyfluoren ≥98%
9-Hydroxyfluoren
Purity: 98+%
CAS No.: 1689-64-1
Molecular Formula: C13H10O
Formula Weight: 182.22
Melting Point: 155-157°
Beilstein Reference: 1869799
Formel:
C₁₃H₁₀O Molecular Weight: 182,22 g/mol Siedepunkt: 320 °C (1013 hPa) Schmelzpunkt: 155…157 °C Dichte: 1,151 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001135 CAS-Nummer: 1689-64-1 EINECS: 216-879-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...