Tropinon ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99%
CAS No.: 532-24-1
Molecular Formula: C8H13NO
Formula Weight: 139.20
Storage Temperature: 2°C to 8°C
Flash Point: 90°(194°F)
Odour: Odourless
Melting Point: 40-43°
Boiling Point: 227°
Beilstein Reference: 2329
MDL No.: MFCD00005549
Formel:
C₈H₁₃NO Siedepunkt: 227 °C Schmelzpunkt: 40…43 °C Flammpunkt: 90 °C (194 °F) |
MDL:
MFCD00005549 CAS-Nummer: 532-24-1 EINECS: 208-530-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...