1,2-Bis(phenylthio)ethan ≥98%

Lieferant: Thermo Fisher Scientific

L02019.09 L02019.18
L02019.09EA 54.6 EUR
L02019.09 L02019.18
1,2-Bis(phenylthio)ethan ≥98%
1,2-Bis(phenylthio)ethan

Purity: 98+% CAS No.: 622-20-8 Molecular Formula: C14H14S2 Formula Weight: 246.39 Melting Point: 68-70° Beilstein Reference: 1877719

Formel: C₁₄H₁₄S₂
Schmelzpunkt: 68…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014085
CAS-Nummer: 622-20-8
EINECS: 210-723-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR