2-Aminobenzimidazol ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Benzimidazol-2-ylamin, 2-Amino-1H-benzimidazol

L02066.06 L02066.14 L02066.22
L02066.06EA 28.7 EUR
L02066.06 L02066.14 L02066.22
2-Aminobenzimidazol ≥97%
2-Aminobenzimidazole

Purity: 97+% CAS No.: 934-32-7 Molecular Formula: C7H7N3 Formula Weight: 133.15 Melting Point: 228-232° Beilstein Reference: 116525

Formel: C₇H₇N₃
Molecular Weight: 133.15 g/mol
Siedepunkt: 381
Schmelzpunkt: 228…232 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005596
CAS-Nummer: 934-32-7
EINECS: 213-280-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR