Cell culture media, Schneider's Insect Medium
Lieferant: Biowest
L0207-500
L0207-500EA
24.9
EUR
L0207-500
Cell culture media, Schneider's Insect Medium
Zellkulturmedien
Schneider’s Insect Medium was developed to support the growth of excised imaginal discs from the fruit fly, Drosophila melanogaster. When supplemented with 5 to 20% heat-inactivated foetal bovine serum Schneider's mediuam has been found to support the rapid growth of both primary and established cultures of cells derived from Drosophila spp. and several other dipterans.