N,N'-Dimethylethylendiamin 95%

Lieferant: Thermo Fisher Scientific
Gefahr

L02204.06 L02204.14
L02204.06EA 46.2 EUR
L02204.06 L02204.14
N,N'-Dimethylethylendiamin 95%
N,N'-Dimethylethylendiamin

Purity: 95% CAS No.: 110-70-3 Molecular Formula: C4H12N2 Formula Weight: 88.15 Flash Point: 28°(82°F) Refractive Index: 1.4300 Odour: Amine-like Boiling Point: 118-119° Beilstein Reference: 878142 UN No.: 2734 MDL No.: MFCD00008290

Formel: C₄H₁₂N₂
Siedepunkt: 118…119 °C
Dichte: 0,818
Flammpunkt: 28 °C (82 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008290
CAS-Nummer: 110-70-3
EINECS: 203-793-3
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR