Isopentylamin 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Isoamylamin, 3-Methylbutylamin, 1-Amino-3-methylbutan

L02260.14
L02260.14EA 45.2 EUR
L02260.14
Isopentylamin 99%
Isopentylamin

Purity: 99% CAS No.: 107-85-7 Molecular Formula: (CH3)2CHCH2CH2NH2 Formula Weight: 87.17 Flash Point: -1°(30°F) Refractive Index: 1.4080 Odour: Amine-like pH: 12.1 Melting Point: -60° Boiling Point: 95-97° Beilstein Reference: 1209230 Merck Reference: 14,5112 UN No.: 1106 MDL No.: MFCD00008203

Formel: C₅H₁₃N
Molecular Weight: 87.16 g/mol
Siedepunkt: 95…97 °C
Schmelzpunkt: –60 °C
Dichte: 0.751
Flammpunkt: –1 °C (30 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008203
CAS-Nummer: 107-85-7
EINECS: 203-526-0
UN: 1106
ADR: 3 II
Merck Index: 13,05128

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR