Triphenylphosphin 99%, Pulver
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L02502.18
L02502.30
L02502.0B
AB172490.0250
L02502.18EA
24.7
EUR
L02502.18
L02502.30
L02502.0B
ABCRAB172490.0250
Triphenylphosphin 99%, Pulver
Triphenylphosphin
Purity: 99%
CAS No.: 603-35-0
Molecular Formula: (C6H5)3P
Formula Weight: 262.29
Flash Point: 181°(358°F)
Odour: Mild
Boiling Point: 360°
Beilstein Reference: 610776
Merck Reference: 14,9743
MDL No.: MFCD00003043
Formel:
C₁₈H₁₅P Siedepunkt: 360 °C Schmelzpunkt: 79…81 °C Dichte: 1,200 Flammpunkt: 181 °C (358 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003043 CAS-Nummer: 603-35-0 EINECS: 210-036-0 Merck Index: 14,9743 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...