2,6-Pyridindimethanol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2,6-Bis(hydroxymethyl)pyridine
L02527.06
L02527.14
L02527.06EA
72.4
EUR
L02527.06
L02527.14
2,6-Pyridindimethanol ≥98%
2,6-Pyridindimethanol
Purity: 98%
CAS No.: 1195-59-1
Molecular Formula: C7H9NO2
Formula Weight: 139.15
Melting Point: 110-114°
Boiling Point: 185°/15mm
MDL No.: MFCD00006351
Formel:
C₇H₉NO₂ Molecular Weight: 139.15 g/mol Siedepunkt: 185 °C (15 mmHg) Schmelzpunkt: 110…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006351 CAS-Nummer: 1195-59-1 EINECS: 214-803-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...