3-(2-Hydroxyethyl)indol ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: β-Indol-3-ylethanol, Indol-3-ethanol, Tryptophol

L02555.03 L02555.06
L02555.03EA 24.8 EUR
L02555.03 L02555.06
3-(2-Hydroxyethyl)indol ≥97%
3-(2-Hydroxyethyl)indole

Purity: 97% CAS No.: 526-55-6 Molecular Formula: C10H11NO Formula Weight: 161.20 Flash Point: 255°(491°F) Melting Point: 56-59° Boiling Point: 174°/2mm Beilstein Reference: 125553 Merck Reference: 14,9798 MDL No.: MFCD00005659

Formel: C₁₀H₁₁NO
Molecular Weight: 161.2 g/mol
Siedepunkt: 174 °C (2 mmHg)
Schmelzpunkt: 56…59 °C
Flammpunkt: 255 °C (491 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005659
CAS-Nummer: 526-55-6
EINECS: 208-393-2
Merck Index: 12,09930

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR