2-Methylpiperidin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98+%
CAS No.: 109-05-7
Molecular Formula: C6H13N
Formula Weight: 99.18
Flash Point: 8°(46°F)
Refractive Index: 1.4470
Odour: Amine-like
Melting Point: -4°
Boiling Point: 118-119°
Beilstein Reference: 79804
UN No.: 2733
MDL No.: MFCD00005982
Formel:
C₆H₁₃N Siedepunkt: 118…119 °C Schmelzpunkt: –4 °C Dichte: 0,842 Flammpunkt: 8 °C (46 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005982 CAS-Nummer: 109-05-7 EINECS: 203-642-1 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...