1-Hexadecen ≥94%
Lieferant: Thermo Fisher Scientific
L02637.AK
L02637.AU
L02637.AKEA
42
EUR
L02637.AK
L02637.AU
1-Hexadecen ≥94%
1-Hexadecen
Purity: 94%
CAS No.: 629-73-2
Molecular Formula: CH3(CH2)13CH=CH2
Formula Weight: 224.43
Flash Point: 132°(269°F)
Refractive Index: 1.4420
Melting Point: 4°
Boiling Point: 273-274°
Beilstein Reference: 1753401
MDL No.: MFCD00008991
Formel:
C₁₆H₃₂ Siedepunkt: 273…274 °C Schmelzpunkt: 4 °C Dichte: 0,781 Flammpunkt: 132 °C (269 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008991 CAS-Nummer: 629-73-2 EINECS: 211-105-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...