2,5-Dibrom-3-methylthiophen ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L02693.06 L02693.14
L02693.06EA 35 EUR
L02693.06 L02693.14
2,5-Dibrom-3-methylthiophen ≥98%
2,5-Dibromo-3-methylthiophene

Purity: 98% CAS No.: 13191-36-1 Molecular Formula: C5H4Br2S Formula Weight: 255.97 Refractive Index: 1.6130 Boiling Point: 226-230° MDL No.: MFCD00015470

Formel: C₅H₄Br₂S
Siedepunkt: 226…230 °C
Dichte: 1,972
Lagertemperatur: Raumtemperatur
MDL: MFCD00015470
CAS-Nummer: 13191-36-1
EINECS: 236-147-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR