3,5-Dimethyl-4-nitrobenzoic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L02704.03
L02704.06
L02704.03EA
67.8
EUR
L02704.03
L02704.06
3,5-Dimethyl-4-nitrobenzoic acid 97%
3,5-Dimethyl-4-nitrobenzoic acid
Purity: 97%
CAS No.: 3095-38-3
Molecular Formula: C9H9NO4
Formula Weight: 195.18
Odour: Odourless
Melting Point: 222-226°
Beilstein Reference: 1965772
MDL No.: MFCD00014699
Formel:
C₉H₉NO₄ Schmelzpunkt: 222…226 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014699 CAS-Nummer: 3095-38-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...