trans-4-(Trifluormethyl)zimtsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L02771.03
L02771.06
L02771.03EA
39.1
EUR
L02771.03
L02771.06
trans-4-(Trifluormethyl)zimtsäure ≥98%
trans-4-(Trifluormethyl)zimtsäure
Purity: 98%
CAS No.: 16642-92-5
Molecular Formula: C10H7F3O2
Formula Weight: 216.16
Odour: Odourless
Melting Point: 230-234°
Beilstein Reference: 2452800
MDL No.: MFCD00002696
Formel:
C₁₀H₇F₃O₂ Schmelzpunkt: 230…234 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002696 CAS-Nummer: 16642-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...