4-Brom-DL-phenylalanin ≥98%

Lieferant: Thermo Fisher Scientific

L02833.03 L02833.06
L02833.03EA 56.1 EUR
L02833.03 L02833.06
4-Brom-DL-phenylalanin ≥98%
4-Brom-DL-phenylalanin

Purity: 98+% CAS No.: 14091-15-7 Molecular Formula: C9H10BrNO2 Formula Weight: 244.09 Odour: Odourless Melting Point: ca 262° dec. Beilstein Reference: 2212158 MDL No.: MFCD00002599

Formel: C₉H₁₀BrNO₂
Molecular Weight: 244.09 g/mol
Schmelzpunkt: ∼ 262 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002599
CAS-Nummer: 14091-15-7
EINECS: 237-941-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR