p-Xylylendiamin 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1,4-Bis(aminomethyl)benzene, 1,4-Phenylenedimethanamine
L02995.04
L02995.09
L02995.18
AB177666.0050
L02995.04EA
43.7
EUR
L02995.04
L02995.09
L02995.18
ABCRAB177666.0050
p-Xylylendiamin 97%
p-Xylylendiamin
Purity: 97%
CAS No.: 539-48-0
Molecular Formula: C8H12N2
Formula Weight: 136.20
Flash Point: 134°(273°F)
Melting Point: 59-63°
Boiling Point: 230°/10mm
Beilstein Reference: 2206190
UN No.: 3259
MDL No.: MFCD00009821
Formel:
C₈H₁₂N₂ Molecular Weight: 136.20 g/mol Siedepunkt: 230 °C (10 mmHg) Schmelzpunkt: 59…63 °C Flammpunkt: 134 °C (273 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009821 CAS-Nummer: 539-48-0 EINECS: 208-719-3 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...