Pentamethylene sulphide 98%

Lieferant: Thermo Fisher Scientific
Gefahr

L03063.03 L03063.06
L03063.03EA 30.1 EUR
L03063.03 L03063.06
Pentamethylene sulphide 98%
Pentamethylene sulphide

Purity: 98% CAS No.: 1613-51-0 Molecular Formula: C5H10S Formula Weight: 102.20 Flash Point: 21°(69°F) Refractive Index: 1.5060 Odour: Unpleasant Boiling Point: 140-142° Beilstein Reference: 102437 UN No.: UN1993 MDL No.: MFCD00006662

Formel: C₅H₁₀S
Siedepunkt: 140…142 °C
Dichte: 0,988
Flammpunkt: 21 °C (69 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006662
CAS-Nummer: 1613-51-0
EINECS: 216-561-1
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR