(Methylthio)acetonitril ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L03066.06
L03066.14
L03066.22
L03066.06EA
30.5
EUR
L03066.06
L03066.14
L03066.22
(Methylthio)acetonitril ≥99%
(Methylthio)acetonitril
Purity: 99%
CAS No.: 35120-10-6
Molecular Formula: C3H5NS
Formula Weight: 87.14
Flash Point: 67°(152°F)
Refractive Index: 1.4820
Boiling Point: 166-167°
Beilstein Reference: 1738015
MDL No.: MFCD00001925
Formel:
C₃H₅NS Siedepunkt: 166…167 °C Dichte: 1,052 Flammpunkt: 67 °C (152 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001925 CAS-Nummer: 35120-10-6 EINECS: 252-383-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...