Tetrabutoxysilan ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 97%
CAS No.: 4766-57-8
Molecular Formula: C16H36O4Si
Formula Weight: 320.55
Flash Point: 135°(275°F)
Refractive Index: 1.4130
Odour: Characteristic
Melting Point:
Formel:
C₁₆H₃₆O₄Si Siedepunkt: 277 °C Schmelzpunkt: < –80 °C Dichte: 0,899 Flammpunkt: 135 °C (275 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009432 CAS-Nummer: 4766-57-8 EINECS: 225-305-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...