2-Hydroxyethyl methyl sulphone 97%

Lieferant: Thermo Fisher Scientific

L03159.06
L03159.06EA 52.9 EUR
L03159.06
2-Hydroxyethyl methyl sulphone 97%
2-Hydroxyethyl methyl sulphone

Purity: 97% CAS No.: 15205-66-0 Molecular Formula: C3H8O3S Formula Weight: 124.16 Flash Point: 110°(230°F) Refractive Index: 1.4790 Melting Point: 27-30° Boiling Point: 148-149°/1mm Beilstein Reference: 1746915 MDL No.: MFCD00014747

Formel: C₃H₈O₃S
Siedepunkt: 148…149 °C (1 mmHg)
Schmelzpunkt: 27…30 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014747
CAS-Nummer: 15205-66-0
EINECS: 239-261-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR