m-Toluenesulphonic acid monohydrate 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L03419.06
L03419.14
L03419.06EA
100
EUR
L03419.06
L03419.14
m-Toluenesulphonic acid monohydrate 97%
m-Toluenesulphonic acid monohydrate
Purity: 97%
CAS No.: 312619-56-0
Molecular Formula: C7H8O3S.H2O
Formula Weight: 190.22 (172.20anhy)
Refractive Index: 1.5270
Boiling Point: 165°/0.1mm
Beilstein Reference: 2208421
UN No.: UN2586
MDL No.: MFCD00167130
Formel:
C₇H₈O₃S·H₂O Siedepunkt: 165 °C (0,1 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00167130 CAS-Nummer: 312619-56-0 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...