4-[Chlor(dimethyl)silyl]butannitril 94%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Chloro(3-cyanopropyl)dimethylsilane
L03607.09
L03607.09EA
48.9
EUR
L03607.09
4-[Chlor(dimethyl)silyl]butannitril 94%
(3-Cyanopropyl)dimethylchlorosilane
Purity: 94%
CAS No.: 18156-15-5
Molecular Formula: C6H12ClNSi
Formula Weight: 161.71
Flash Point: 85°(185°F)
Refractive Index: 1.4460
Boiling Point: 108-109°/15mm
Beilstein Reference: 2235062
UN No.: UN2987
MDL No.: MFCD00001969
Formel:
C₆H₁₂ClNSi Molecular Weight: 161,70 g/mol Siedepunkt: 108…109 °C (15 mmHg) Dichte: 0,990 Flammpunkt: 85 °C (185 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001969 CAS-Nummer: 18156-15-5 EINECS: 242-039-8 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...