Ethylammoniumchlorid 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethylamin Hydrochlorid, Aminoethan Hydrochlorid

L03624.30 L03624.0B
L03624.30EA 44.5 EUR
L03624.30 L03624.0B
Ethylammoniumchlorid 98%
Ethylammonium chloride

Bitte beachten Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Formel: C₂H₈ClN
Molecular Weight: 81.54 g/mol
Schmelzpunkt: 95…100 °C
Dichte: 1.220
Lagertemperatur: Raumtemperatur
MDL: MFCD00012885
CAS-Nummer: 557-66-4
EINECS: 209-182-8
Merck Index: 14,3762

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR