Cyclohexyl-methylsulfid ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

L03712.06 L03712.14
L03712.06EA 46.6 EUR
L03712.06 L03712.14
Cyclohexyl-methylsulfid ≥97%
Cyclohexyl-methylsulfid

Purity: 97% CAS No.: 7133-37-1 Molecular Formula: C7H14S Formula Weight: 130.25 Flash Point: 61°(142°F) Refractive Index: 1.4940 Odour: Unpleasant Melting Point: -80° Boiling Point: 89-90°/35mm Beilstein Reference: 1919309 UN No.: 3334 MDL No.: MFCD00039460

Formel: C₇H₁₄S
Siedepunkt: 89…90 °C (35 mmHg)
Schmelzpunkt: –80 °C
Dichte: 0,944
Flammpunkt: 61 °C (142 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00039460
CAS-Nummer: 7133-37-1
EINECS: 230-431-1
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR