Methyl-4-chlorbutyrat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 4-chlorobutyrate

L03734.14 L03734.22
L03734.14EA 23.6 EUR
L03734.14 L03734.22
Methyl-4-chlorbutyrat ≥98%
Methyl-4-chlorbutyrat

Purity: 98% CAS No.: 3153-37-5 Molecular Formula: Cl(CH2)3CO2CH3 Formula Weight: 136.58 Flash Point: 59°(138°F) Refractive Index: 1.4320 Boiling Point: 175-176° Beilstein Reference: 1745117 UN No.: 3272 MDL No.: MFCD00001003

Formel: C₅H₉ClO₂
Molecular Weight: 136.58 g/mol
Siedepunkt: 175…176 °C
Dichte: 1.125
Flammpunkt: 59 °C (138 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001003
CAS-Nummer: 3153-37-5
EINECS: 221-592-9
UN: 3272
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR