(Benzylthio)acetone 98%

Lieferant: Thermo Scientific

L03814.14
L03814.14EA 119 EUR
L03814.14
(Benzylthio)acetone 98%
(Benzylthio)acetone

Purity: 98% CAS No.: 10230-69-0 Molecular Formula: C10H12OS Formula Weight: 180.27 Flash Point: 100°(212°F) Refractive Index: 1.5580 Boiling Point: 155-156°/17mm Beilstein Reference: 1866268 MDL No.: MFCD00026241

Formel: C₁₀H₁₂OS
Molecular Weight: 180,27 g/mol
Siedepunkt: 155…156 °C (17 mmHg)
Dichte: 1,090
Flammpunkt: >100 °C (212 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00026241
CAS-Nummer: 10230-69-0
EINECS: 233-552-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR