L(+)-Norleucin ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

L03913.03 L03913.06
L03913.03EA 39.5 EUR
L03913.03 L03913.06
L(+)-Norleucin ≥99%
L(+)-Norleucin

Purity: 99% CAS No.: 327-57-1 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: 300° Beilstein Reference: 1721750 Merck Reference: 14,6706 Optical Rotation: +23° (c=5 in 5N HCl) MDL No.: MFCD00064423

Formel: C₆H₁₃NO₂
Siedepunkt: 246 °C (1013 hPa)
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064423
CAS-Nummer: 327-57-1
EINECS: 206-321-4
Merck Index: 14,6706

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR