Acetophenonazin ≥97%

Lieferant: Thermo Scientific

Synonyme: 1,2-Bis(1-phenylethylidene)hydrazine

L03924.06
L03924.06EA 160 EUR
L03924.06
Acetophenonazin ≥97%
Acetophenonazin

Purity: 97% CAS No.: 729-43-1 Molecular Formula: C16H16N2 Formula Weight: 236.32 Melting Point: 120-124° Beilstein Reference: 1912471

Formel: C₁₆H₁₆N₂
Molecular Weight: 236,32 g/mol
Schmelzpunkt: 120…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015011
CAS-Nummer: 729-43-1
EINECS: 211-979-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR