9-Fluorenone oxime ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
9H-Fluoren-9-one oxime
L03950.06
L03950.06EA
34.7
EUR
L03950.06
9-Fluorenone oxime ≥98%
9-Fluorenone oxime
Purity: 98+%
CAS No.: 2157-52-0
Molecular Formula: C13H9NO
Formula Weight: 195.22
Melting Point: 193-194°
Beilstein Reference: 1871046
MDL No.: MFCD00016356
Formel:
C₁₃H₉NO Molecular Weight: 195.22 g/mol Schmelzpunkt: 193…194 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016356 CAS-Nummer: 2157-52-0 EINECS: 218-471-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...