Dimethoxy(diphenyl)silan ≥97%
Lieferant: Thermo Fisher Scientific
L04031.18
L04031.30
L04031.0B
L04031.18EA
31.9
EUR
L04031.18
L04031.30
L04031.0B
Dimethoxy(diphenyl)silan ≥97%
Dimethoxy(diphenyl)silan
Purity: 97%
CAS No.: 6843-66-9
Molecular Formula: C14H16O2Si
Formula Weight: 244.37
Flash Point: 121°(249°F)
Refractive Index: 1.5420
Boiling Point: 160-162°/15mm
Beilstein Reference: 2940458
MDL No.: MFCD00025690
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...