Dimethoxy(diphenyl)silan ≥97%
Lieferant: Thermo Fisher Scientific
L04031.18 L04031.30 L04031.0B
L04031.18EA 31.9 EUR
L04031.18 L04031.30 L04031.0B
Dimethoxy(diphenyl)silan ≥97%
Dimethoxy(diphenyl)silan
Purity: 97% CAS No.: 6843-66-9 Molecular Formula: C14H16O2Si Formula Weight: 244.37 Flash Point: 121°(249°F) Refractive Index: 1.5420 Boiling Point: 160-162°/15mm Beilstein Reference: 2940458 MDL No.: MFCD00025690