1,2-Bis(phenylsulphonyl)ethane ≥98%

Lieferant: Thermo Fisher Scientific

L04083.06 L04083.14
L04083.06EA 59.6 EUR
L04083.06 L04083.14
1,2-Bis(phenylsulphonyl)ethane ≥98%
1,2-Bis(phenylsulphonyl)ethane

Purity: 98+% CAS No.: 599-94-0 Molecular Formula: C14H14O4S2 Formula Weight: 310.39 Melting Point: 179-180° Beilstein Reference: 1888062 MDL No.: MFCD00041256

Formel: C₁₄H₁₄O₄S₂
Molecular Weight: 310.39 g/mol
Schmelzpunkt: 179…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041256
CAS-Nummer: 599-94-0
EINECS: 209-979-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR