3-Fluorbenzalchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: α,α-Dichlor-3-fluortoluol

L04121.06
L04121.06EA 74.6 EUR
L04121.06
3-Fluorbenzalchlorid ≥98%
3-Fluorbenzalchlorid

Purity: 98% CAS No.: 402-64-2 Molecular Formula: C7H5Cl2F Formula Weight: 179.02 Refractive Index: 1.5275 Boiling Point: 195-197° UN No.: 3265 MDL No.: MFCD00037112

Formel: C₇H₅Cl₂F
Molecular Weight: 179,02 g/mol
Siedepunkt: 195…197 °C
Dichte: 1,36 g/cm³ (16 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00037112
CAS-Nummer: 402-64-2
EINECS: 206-952-5
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR