Chlor(methyl)diphenylsilan ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97%
CAS No.: 144-79-6
Molecular Formula: C13H13ClSi
Formula Weight: 232.79
Flash Point: 141°(285°F)
Refractive Index: 1.5740
Odour: Irritating
Boiling Point: 294-295°
Beilstein Reference: 2937445
UN No.: 2987
MDL No.: MFCD00000498
Formel:
C₁₃H₁₃ClSi Siedepunkt: 294…295 °C Dichte: 1,104 Flammpunkt: 141 °C (285 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000498 CAS-Nummer: 144-79-6 EINECS: 205-639-0 UN: 2987 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...