Sulphamerazine ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-Amino-N-(4-methylpyrimidin-2-yl)benzenesulphonamide

L04194.14 L04194.22
L04194.14EA 23.2 EUR
L04194.14 L04194.22
Sulphamerazine ≥98%
Sulphamerazine

Purity: 98+% CAS No.: 127-79-7 Molecular Formula: C11H12N4O2S Formula Weight: 264.30 Melting Point: 234-238° Beilstein Reference: 249133 MDL No.: MFCD00023212

Formel: C₁₁H₁₂N₄O₂S
Schmelzpunkt: 234…238 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00023212
CAS-Nummer: 127-79-7
EINECS: 204-866-2
Merck Index: 14,8913

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR