Bis(4-hydroxyphenyl)sulfid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

L04308.22 L04308.36
L04308.22EA 48.5 EUR
L04308.22 L04308.36
Bis(4-hydroxyphenyl)sulfid ≥98%
Bis(4-hydroxyphenyl)sulfid

Purity: 98+% CAS No.: 2664-63-3 Molecular Formula: C12H10O2S Formula Weight: 218.27 Melting Point: 151-155° Beilstein Reference: 2050739 UN No.: 3261 MDL No.: MFCD00002349

Formel: C₁₂H₁₀O₂S
Schmelzpunkt: 151…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002349
CAS-Nummer: 2664-63-3
EINECS: 220-197-9
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR