(1-Tetradecyl)triphenylphosphoniumbromid ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Triphenyl(tetradecyl)phosphonium bromide, Tetradecyltriphenylphosphonium bromide

L04311.14 L04311.22
L04311.14EA 124 EUR
L04311.14 L04311.22
(1-Tetradecyl)triphenylphosphoniumbromid ≥97%
(1-Tetradecyl)triphenylphosphoniumbromid

Purity: 97% CAS No.: 25791-20-2 Molecular Formula: CH3(CH2)13P(C6H5)3Br Formula Weight: 539.59 Melting Point: 87-89° Beilstein Reference: 3582243

Formel: H₃C(CH₂)₁₃P(C₆H₅)₃Br
Schmelzpunkt: 87…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051857
CAS-Nummer: 25791-20-2
EINECS: 247-267-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR