(1-Tetradecyl)triphenylphosphoniumbromid ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Triphenyl(tetradecyl)phosphonium bromide, Tetradecyltriphenylphosphonium bromide
L04311.14
L04311.22
L04311.14EA
124
EUR
L04311.14
L04311.22
(1-Tetradecyl)triphenylphosphoniumbromid ≥97%
(1-Tetradecyl)triphenylphosphoniumbromid
Purity: 97%
CAS No.: 25791-20-2
Molecular Formula: CH3(CH2)13P(C6H5)3Br
Formula Weight: 539.59
Melting Point: 87-89°
Beilstein Reference: 3582243
Formel:
H₃C(CH₂)₁₃P(C₆H₅)₃Br Schmelzpunkt: 87…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051857 CAS-Nummer: 25791-20-2 EINECS: 247-267-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...