4,4'-cis-Stilbenedicarboxylic acid 95%

Lieferant: Thermo Scientific
Achtung

L04364.03
L04364.03EA 52.4 EUR
L04364.03
4,4'-cis-Stilbenedicarboxylic acid 95%
4,4'-cis-Stilbenedicarboxylic acid

Purity: 95% CAS No.: 133005-88-6 Molecular Formula: C16H12O4 Formula Weight: 268.27 Melting Point: 300° Beilstein Reference: 1978660 MDL No.: MFCD00013994

Formel: C₁₆H₁₂O₄
Molecular Weight: 268,27 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013994
CAS-Nummer: 133005-88-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR